Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYBC1 Antibody (OTI3B5), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272205AF488
Description
CYBC1 Monoclonal specifically detects CYBC1 in Human samples. It is validated for Western Blot.Specifications
| CYBC1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYBC1 | |
| Full length recombinant protein of human C17orf62 (NP_001028218) produced in HEK293T cells. | |
| 0.1 mL | |
| 79415.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3B5 | |
| Western Blot | |
| C17orf62 chromosome 17 open reading frame 62 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction