Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYBC1 Antibody (OTI3B5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272205AF532
Description
CYBC1 Monoclonal specifically detects CYBC1 in Human samples. It is validated for Western Blot.Specifications
CYBC1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CYBC1 | |
Full length recombinant protein of human C17orf62 (NP_001028218) produced in HEK293T cells. | |
0.1 mL | |
79415.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3B5 | |
Western Blot | |
C17orf62 chromosome 17 open reading frame 62 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction