Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cybr Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991917
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1917 25 μL
50-199-1916 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1917 Supplier Rockland Immunochemicals Supplier No. 600401A23S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Cybr. A BLAST analysis was used to suggest cross-reactivity with Cybr from mouse and rat based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.

Specifications

Antigen Cybr
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CYTIP
Gene Accession No. Q5I0L6, Q91VY6
Gene Alias A130053M09Rik; AI462064; B3-1; BOS_1774; C80816; CASP; Cbp; Cbp HE; CYBR; CYTHIP; Cytip; cytohesin 1 interacting protein; cytohesin 1 interacting protein-like; Cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; Cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; Cytohesin-interacting protein; HE; LOC100354602; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; Pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; PSCDBP; Sec7; tamalin
Gene Symbols CYTIP
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227929, 311047
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.