Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYBRD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 267351AP150UL

Description
This gene is a member of the cytochrome b(561) family that encodes an iron-regulated protein. It highly expressed in the duodenal brush border membrane. It has ferric reductase activity and is believed to play a physiological role in dietary iron absorption.Specifications
CYBRD1 | |
Polyclonal | |
Unconjugated | |
CYBRD1 | |
CYBRD1, cytochrome b reductase 1, DCYTB, Duodenal cytochrome b, Ferric chelate reductase 3, FRRS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79901 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.63 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53TN4 | |
CYBRD1 | |
CYBRD1 Fusion Protein Ag24968 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction