Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclic adenosine monophosphate (cAMP) Monoclonal Antibody (ABM486), Biotin, Abbomax

Catalog No. 502203847
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3847 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3847 Supplier Abbomax Inc Supplier No. 500953B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: C-AMP-BSA Cellular location: Cytoplasmic.

Cyclic adenosine monophosphate (cAMP) Monoclonal antibody specifically detects Cyclic adenosine monophosphate (cAMP) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Cyclic adenosine monophosphate (cAMP)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ABM486
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP
Host Species Mouse
Immunogen A chemically linked 3, 5-Cyclic Adenosine Monophosphate (cAMP)
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.