Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cyclin A Ab-6, Mouse Monoclonal Antibody

Catalog No. MS1061R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1061R7 7 mL Prediluted
MS1061S0 100 μL Concentrated
MS1061S1 500 μL Concentrated
MS1061S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1061R7 Supplier Epredia™ Supplier No. MS1061R7

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.

Host Species: Mouse

Clone: 6E6

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant N-terminal fragment of human cyclin A protein

Molecular Weight: 60 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin-fixed only)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cyclin A Ab-6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6E6
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant N-terminal fragment of human cyclin A protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.