Learn More
Description
Host Species: Mouse
Clone: 6E6
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant N-terminal fragment of human cyclin A protein
Molecular Weight: 60 kDa
Positive Control: HeLa cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin-fixed only)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cyclin A Ab-6 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 6E6 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant N-terminal fragment of human cyclin A protein |
| Purification Method | HeLa cells, tonsil |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.