Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109491 Supplier Invitrogen™ Supplier No. PA5109491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: METGFPAIMY PGSFIGGWGE EYLSWEGPGL PDFVFQQPVE SEAMHCSNPK SGVVLATVAR GPDACQILTR APLGQDPPQR TVLGLLTANG QYRRTCGQGI TRIRCYSGSE NAFPPAGKKA LPDCGVQEPP KQGFDIYMDE LEQGDRDSCS VREGMAFEDV YEVDTGTLKS DLHFLLDFNT VSPMLVDSSL LSQSEDISSL GTDVINVTEY AEEIYQYLRE AEIRHRPKAH.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCNA1
Gene Accession No. P78396, Q6AY13
Gene Alias CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34
Gene Symbols CCNA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human CCNA1 (NP_001104515.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295052, 8900
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.