Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1 Antibody (XLA1-1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002624AF700
Description
Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.Specifications
Cyclin A1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCNA1 | |
Xenopus laevis cyclin A | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
8900 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
XLA1-1 | |
Western Blot | |
cyclin A1, cyclin-A1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction