Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1 Antibody (XLA1-1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308717IR
Description
Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Xenopus samples. It is validated for Western BlotSpecifications
Cyclin A1 | |
Monoclonal | |
DyLight 755 | |
cyclin A1, cyclin-A1 | |
Recombinant full-length Xenopus laevis Cyclin A1 protein (Uniprot: P18606) | |
100 μg | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
8900 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot | |
XLA1-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction