Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1/Cyclin A2 Antibody (SR1909), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Cyclin A1/Cyclin A2 |
---|---|
Clone | SR1909 |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
Cyclin A1/Cyclin A2 Monoclonal antibody specifically detects Cyclin A1/Cyclin A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Cyclin A1/Cyclin A2 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
IgG | |
Affinity purified |
SR1909 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
A synthesized peptide derived from human Cyclin A1/Cyclin A2 (Uniprot #: P20248, P78396) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title