Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1 Monoclonal Antibody (3F6B2C2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA529091
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CCNA1/Cyclin-A1.
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.
Specifications
Cyclin A1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P78396 | |
CCNA1 | |
Recombinant Human CCNA1 protein (Met1-Gln465) | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Western Blot | |
3F6B2C2 | |
Unconjugated | |
CCNA1 | |
CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34 | |
Mouse | |
Protein A | |
RUO | |
8900 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction