Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1 Mouse anti-Xenopus, Clone: XLA1-1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | Cyclin A1 |
|---|---|
| Clone | XLA1-1 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml |
| Applications | Western Blot |
Description
Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.Specifications
| Cyclin A1 | |
| 0.2 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 8900 | |
| IgG1 κ | |
| Protein A or G purified |
| XLA1-1 | |
| Western Blot 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Xenopus species | |
| cyclin A1, cyclin-A1 | |
| Recombinant full-length Xenopus laevis Cyclin A1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title