Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Mouse anti-Xenopus, Clone: XLA1-3, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Cyclin A1
Clone XLA1-3
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122960
SDP
View Documents
Novus Biologicals
NBP30774320UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.
Specifications

Specifications

Cyclin A1
0.2 mg/ml
Western Blot
Unconjugated
Mouse
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
10 mM PBS with 0.05% BSA
8900
IgG1 κ
Protein A or G purified
XLA1-3
Western Blot 1-2 ug/ml
Monoclonal
Purified
RUO
Xenopus species
cyclin A1, cyclin-A1
Recombinant full-length Xenopus laevis Cyclin A1 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.