Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | Cyclin A1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5122144
|
Invitrogen™
PA5122144 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5122143
|
Invitrogen™
PA5122143 |
100 μL |
Each for $511.50
|
|
|||||
Description
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.Specifications
Cyclin A1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P78396, Q61456, Q6AY13 | |
12427, 295052, 8900 | |
Recombinant CCNA1 (Ser183-Tyr430) expressed in E. coli. | |
Primary | |
-20°C | |
CCNA1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
CCNA1; CT146; cyclin A1; cyclin-A1; testicular tissue protein Li 34 | |
CCNA1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title