Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986763
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6763 Supplier Bioss Supplier No. BS5739R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk′s). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.

Specifications

Antigen Cyclin A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCNA1
Gene Accession No. P78396
Gene Alias CCNA1; CT146; cyclin A1; Cyclin-A1; testicular tissue protein Li 34
Gene Symbols CCNA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Cyclin A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8900
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.