Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A2 Antibody (46B11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002615H
Description
Cyclin A2 Monoclonal specifically detects Cyclin A2 in Xenopus samples. It is validated for Western Blot.Specifications
Cyclin A2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CCNA2 | |
SDS derived male Xenopus laevis cyclin A2 fusion protein. | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
890 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
46B11 | |
Western Blot | |
CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction