Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin A2 Recombinant Rabbit Monoclonal Antibody (3E9Z3)
GREENER_CHOICE

Catalog No. PIMA535218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535218 Supplier Invitrogen™ Supplier No. MA535218
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: TREAGSALLA LQQTALQEDQ ENINPEKAAP VQQPRTRAAL AVLKSGNPRG LAQQQRPKTR RVAPLKDLPV NDEHVTVPPW KANSKQPAFT IHVDEAEKEA QKKPAESQKI EREDALAFNS AISLPGPRKP LVPLDYPMDG SFESPHTMDM SIILEDEKPV SVNEVPDYHE DIHTYLREME VKCKPKVGYM KKQPDITNSM.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3E9Z3
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCNA2
Gene Accession No. P20248, P51943
Gene Alias AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg
Gene Symbols CCNA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 11-210 of human Cyclin A2 (P20248).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114494, 12428, 890
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.