Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin A2 Monoclonal Antibody (E23.1)
GREENER_CHOICE

Catalog No. PIMA1154 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1154 Supplier Invitrogen™ Supplier No. MA1154
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-154 detects Cyclin A in mammalian samples and has been successfully used in Immunohistochemistry (Frozen and Paraffin), Immunofluorescence, Immunoprecipitation and Western Blot procedures. Immunoprecipitation and Western Blot analysis with MA1-154 show the accumulation of a prominent band at ~50 kDa in camptothecin and hydroxyurea treated cells. In Immunofluorescence applications, MA1-154 shows nuclear translocation of Cyclin A in G2/M arrested cells as a result of camptothecin treatment.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A2
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone E23.1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene CCNA2
Gene Accession No. P20248, P30274, P51943
Gene Alias AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg
Gene Symbols CCNA2
Host Species Mouse
Immunogen Bovine Cyclin A fusion protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114494, 12428, 281667, 890
Target Species Human, Mouse, Rat, Monkey, Bovine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.