Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin A2 Mouse anti-Human, Clone: 1C6C11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Cyclin A2 |
---|---|
Clone | 1C6C11 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6820
![]() ![]() |
Proteintech Group Inc
663911IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.Specifications
Cyclin A2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CCN1, CCNA, CCNA2, Cyclin A, Cyclin A2 | |
CCNA2 | |
IgG2a | |
Protein A | |
Antibody |
1C6C11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P20248 | |
890 | |
Cyclin A2 Fusion Protein Ag12765 | |
Primary | |
-20°C | |
CCNA2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title