Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A2 Mouse anti-Human, Clone: 2E9, Bioss
SDP

Catalog No. 501989541
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9541 Supplier Bioss Supplier No. BSM52043M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.

Specifications

Antigen Cyclin A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2E9
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene CCNA2
Gene Accession No. P20248
Gene Alias AA408589; BOS_6308; CCN1; Ccn-1; Ccna; Ccna2; Cyca; Cyca2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); Cyclin-A; Cyclin-A2
Gene Symbols CCNA2
Host Species Mouse
Immunogen Recombinant human Cyclin A2 protein, around N-terminal 150aa.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 890
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.