Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin A2 Mouse anti-Human, Clone: CCNA2/2333, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Cyclin A2
Clone CCNA2/2333
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122410
SDP
View Documents
Novus Biologicals
NBP30746820UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Cyclin A2 Monoclonal specifically detects Cyclin A2 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cyclin A2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
10 mM PBS with 0.05% BSA
890
IgG2a κ
Protein A or G purified
CCNA2/2333
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2
Full length recombinant Cyclin A2 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.