Learn More
Invitrogen™ Cyclin A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534682
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, IMR32, SK-N-AS, HeLa(100 μg/mL Bleomycin treatment for 24 hr), Neuro2A, PC-12, PC3. Predicted reactivity: Rat (80%), Pig (89%), Rabbit (84%), Rhesus Monkey (98%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
Specifications
Cyclin A2 | |
Polyclonal | |
Unconjugated | |
CCNA2 | |
AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114494, 12428, 890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.73 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P20248, P51943 | |
CCNA2 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 1 and 190 of Cyclin A2. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.