Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B1 Antibody (2061A), Alexa Fluor 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB6000AF750
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Cyclin B1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CCNB1 | |
Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635] | |
Primary | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry | |
2061A | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
Rabbit | |
0.1 mL | |
891 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction