Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B1 Antibody (2061A), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB6000H
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cyclin B1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CCNB1 | |
Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635] | |
100 μL | |
Cancer, Cell Biology, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
891 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
2061A | |
Western Blot, Immunohistochemistry | |
CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction