Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B1 Antibody (V92.1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002648MFV610
Description
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human, Mouse, Hamster samples. It is validated for Flow CytometrySpecifications
Cyclin B1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Hamster | |
Purified |
Flow Cytometry | |
V92.1 | |
Flow Cytometry | |
CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
Hamster Cyclin B1 | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors | |
891 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction