Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin B1 Monoclonal Antibody (1B10)
GREENER_CHOICE

Catalog No. PIMA515706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515706 Supplier Thermo Scientific Supplier No. MA515706
Add to Cart
Add to Cart

Cyclin B1 Monoclonal Antibody (1B10)

Cyclin B1 Monoclonal Antibody for Western Blot, IHC, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Ccnb1
Gene Accession No. P14635, P30277
Gene Alias Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023
Gene Symbols Ccnb1
Host Species Mouse
Immunogen Purified recombinant fragment of human CCNB1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25203, 891
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.