Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B2 Antibody (X29.2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308843AF700
Description
Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Human, Mouse, Rat, Xenopus samples. It is validated for ELISASpecifications
Cyclin B2 | |
Monoclonal | |
Alexa Fluor 700 | |
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
Recombinant full-length Xenopus laevis Cyclin B2 protein (Uniprot: O95067) | |
100 μg | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
9133 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
X29.2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction