Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B2 Antibody (X29.2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002649MFV450
Description
Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Mammalia, Xenopus samples. It is validated for Flow CytometrySpecifications
Cyclin B2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mammalia, Xenopus species | |
Purified |
Flow Cytometry | |
X29.2 | |
Flow Cytometry | |
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
Xenopus laevis cyclin B2 | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
9133 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction