Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin B2 Monoclonal Antibody (X29.2)
GREENER_CHOICE

Catalog No. PIMA1156
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1156 Supplier Invitrogen™ Supplier No. MA1156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-156 detects Cyclin B2 in mammalian samples. MA1-156 has been successfully used in Immunofluorescence, Immunoprecipitation, IHC (P) and Western Blot procedures. Immunoprecipitation and Western Blot analysis with MA1-156 show the accumulation of a prominent band at ~51 kDa in camptothecin and hydroxyurea treated cells. MA1-156 also detects additional unknown band at ~80 kDa. In Immunofluorescence applications, MA1-156 shows cyclin B2 staining consistent with the Golgi region, whereas MA1-155 shows cyclin B1 co-localization with microtubules. MA1-156 reacts with Cyclin B2 from Xenopus laevis and mammalian sources. In Western blot applications, X29.2 also cross reacts with Cyclin B1.

Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone X29.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Ccnb2
Gene Accession No. O95067, P13351, P30276
Gene Alias C-B2; CCNB2; ccnb2.S; Cycb2; cyclin B2; cyclin B2 S homeolog; cyclinb2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2; HsT17299; XELAEV_18020679mg
Gene Symbols Ccnb2, ccnb2.S
Host Species Mouse
Immunogen Xenopus laevis cyclin B2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12442, 397743, 9133
Target Species Human, Mouse, Monkey, Frog
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.