Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin B2 Mouse anti-Human, Mouse, Rat, Xenopus, Clone: X29.2, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cyclin B2 |
---|---|
Clone | X29.2 |
Concentration | 0.2 mg/ml |
Dilution | ELISA |
Applications | ELISA |
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Human, Mouse, Rat, Xenopus samples. It is validated for ELISA.Specifications
Cyclin B2 | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
9133 | |
IgG1 κ | |
Protein A or G purified |
X29.2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Xenopus species | |
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
Recombinant full-length Xenopus laevis Cyclin B2 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title