Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529233 Supplier Invitrogen™ Supplier No. PA529233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299. Predicted reactivity: Mouse (95%), Rat (95%), Dog (95%), Pig (96%), Chicken (81%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Ccnb2
Gene Accession No. O95067
Gene Alias C-B2; CCNB2; ccnb2.S; Cycb2; cyclin B2; cyclin B2 S homeolog; cyclinb2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2; HsT17299; XELAEV_18020679mg
Gene Symbols Ccnb2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cyclin B2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363088, 9133
Target Species Human, Monkey, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.