Learn More
Invitrogen™ Cyclin B2 Recombinant Rabbit Monoclonal Antibody (1L9B9)

Description
Positive test controls include: Rat testis. Immunogen sequence: MALLRRPTVS SDLENIDTGV NSKVKSHVTI RRTVLEEIGN RVTTRAAQVA KKAQNTKVPV QPTKTTNVNK QLKPTASVKP VQMEKLAPKG PSPTPEDVSM.
Specifications
Specifications
| Antigen | Cyclin B2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 1L9B9 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Ccnb2 |
| Gene Accession No. | O95067, P30276 |
| Gene Alias | C-B2; CCNB2; ccnb2.S; Cycb2; cyclin B2; cyclin B2 S homeolog; cyclinb2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2; HsT17299; XELAEV_18020679mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.