Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin D1 Antibody (CL12323), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Cyclin D1 |
---|---|
Clone | CL12323 |
Dilution | Immunocytochemistry/Immunofluorescence 2-10 ug/ml |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for ImmunofluorescenceSpecifications
Cyclin D1 | |
Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
This antibody was developed against a Synthetic Peptide of P24385. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL12323 | |
Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
PBS, pH 7.2, 40% glycerol | |
595 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title