Learn More
Cyclin D1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Cyclin D1 |
---|---|
Concentration | 267 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8541
![]() ![]() |
Proteintech Group Inc
269391AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The Cyclin D1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Cyclin D1. This antibody recognizes human antigen. The Cyclin D1 antibody has been validated for the following applications: FC, IF, WB, ELISA analysis.
Specifications
Cyclin D1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
IgG |
267 μg/mL | |
Polyclonal | |
Rabbit | |
595 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.