Learn More
Description
Specifications
Specifications
| Antigen | Cyclin D2 |
| Applications | ELISA, Peptide Array |
| Classification | Monoclonal |
| Clone | CCND2/2620 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
