Learn More
Invitrogen™ Cyclin D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595922
Description
Immunogen sequence: MELLCHEVDP VRRAVRDRNL LRDDRVLQNL LTIEERYLPQ CSYFKCVQKD IQPYMRRMVA TWMLEVCEEQ KCEEEVFPLA MNYLDRFLAG VPTPKSHLQL LGAVCMFLAS KLKETSPLTA EKLCIYTDNS IKPQELLEWE LVVLGKLKWN LAAVTPHDFI EHILRKLPQQ REKLSLIRKH AQTFIALCAT DFKFAMYPPS MIATGSVGAA ICGLQQDEEV SSLTCDALTE LLAKITNTDV DCLKACQEQI EAVLLNSLQQ YRQDQRDGSK SEDELDQAST PTDVRDIDL; Positive Samples: HeLa, NIH/3T3, Mouse lung, Mouse spleen, Rat lung; Cellular Location: Cytoplasm, Membrane, Nucleus.
Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
Specifications
Cyclin D2 | |
Polyclonal | |
Unconjugated | |
CCND2 | |
2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12444, 64033, 894 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.11 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30279, P30280, Q04827 | |
CCND2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-289 of human Cyclin D2 (NP_0017501). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.