Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclin D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597550
Description
Cyclin D2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
Specifications
Cyclin D2 | |
Polyclonal | |
Unconjugated | |
CCND2 | |
2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
894 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30279 | |
CCND2 | |
Recombinant Human G1/S-specific cyclin-D2 protein (1-289AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction