Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin D3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298894MFV500
Description
Cyclin D3 Polyclonal antibody specifically detects Cyclin D3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Cyclin D3 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cyclin D3. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
896 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction