Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Mouse anti-Human,Canine,Rat, Unconjugated, Clone: DCS-22, MilliporeSigma™

Catalog No. MABE19171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE19171MI 100 μL
MABE19172MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE19171MI Supplier MilliporeSigma Supplier No. MABE1917100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human,Canine,Rat samples. It is validated for Dot Blot, Enzyme Immunoassay (ELISA), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin D3
Applications Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-22
Conjugate Unconjugated
Gene Accession No. NP_001751.1
Gene Alias G1/S-specific cyclin-D3
Gene Symbols CCND3
Host Species Mouse
Immunogen His-tagged, full-length recombinant human Cyclin D3.
Molecular Weight of Antigen ∼33 kDa observed; 32.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone DCS-22 is a mouse monoclonal antibody that detects human Cyclin D3. It targets an epitope within 20 amino acids from the C-terminal region.
Target Species Human, Canine, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.