Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992058
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2058 100 μg
50-199-2059 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2058 Supplier Rockland Immunochemicals Supplier No. 600401GN2
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Cyclin D3 affinity-purified antibody is directed against Cyclin D3 protein. A BLAST analysis was used to suggest reactivity with this protein from mouse, human, and bovine sources based on 100% homology for the immunogen sequence. Cross reactivity with other sources has not been determined. No reactivity was observed against Cyclin D2.

The immunogen for the anti-h-Cyclin D3 was a recombinant full length Cyclin D3 protein recognizing a 34 kD protein. Cyclin D3 is a G1 cyclin closely related to Cyclins D1 and D2 phase progression. Like Cyclin D1 and Cyclin D2, it is activated by the same mechanisms and can phosphorylate pRB when associated with cdk4 and cdk6.

Specifications

Antigen Cyclin D3
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CCND3
Gene Accession No. P30282
Gene Alias 9230106B05Rik; AA682053; AL024085; AW146355; C78795; CCND 3; CCND3; CGD3; cyclin D3; CyclinD3; Cyl-3; D3 type cyclin; D3-type cyclin; G1/S-specific cyclin-D3; OTTHUMP00000223415; RP5-973N23.3
Gene Symbols CCND3
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12445
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.