Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Rabbit anti-Human, Clone: 4A8, Bioss
SDP

Catalog No. 501989545
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9545 Supplier Bioss Supplier No. BSM52047R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

The immunogen for the anti-h-Cyclin D3 was a recombinant full length Cyclin D3 protein recognizing a 34 kD protein. Cyclin D3 is a G1 cyclin closely related to Cyclins D1 and D2 phase progression. Like Cyclin D1 and Cyclin D2, it is activated by the same mechanisms and can phosphorylate pRB when associated with cdk4 and cdk6.

Specifications

Antigen Cyclin D3
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4A8
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene CCND3
Gene Accession No. P30281
Gene Alias 9230106B05Rik; AA682053; AL024085; AW146355; C78795; CCND 3; CCND3; CGD3; cyclin D3; CyclinD3; Cyl-3; D3 type cyclin; D3-type cyclin; G1/S-specific cyclin-D3; OTTHUMP00000223415; RP5-973N23.3
Gene Symbols CCND3
Host Species Rabbit
Immunogen Recombinant human Cyclin D3 protein, around C-terminal 200aa.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 896
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.