Learn More
Description
Host Species: Mouse
Clone: 13A3
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human cyclin E protein
Molecular Weight: Doublet of 50 kDa and a single band of 42 kDa
Positive Control: SKBR3 and LS174T cells; Breast CA or placenta
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cyclin E Ab-5 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 13A3 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human cyclin E protein |
| Purification Method | SKBR3 and LS174T cells, breast CA or placenta |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.