Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Cyclin E1
Concentration 1 mg/mL
Dilution Western Blot 1:500 -1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018176
SDP
View Documents
Novus Biologicals
NB500129SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB500129
SDP
View Documents
Novus Biologicals
NB500129
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Cyclin E1 Polyclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cyclin E1
Western Blot 1:500 -1:1000
Polyclonal
Rabbit
Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
PBS with 0.02% Sodium Azide
898
Partial residue sequence of Cyclin E1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
43 kDa
1 mg/mL
Western Blot
Unconjugated
RUO
Human
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
CCNE1
IgG
Affinity Purified
Reacts with residues 43-52 [GSQPWDNNAVC] of Cyclin E1.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.