Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin E1 Antibody (CCNE1/2460), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308805MFV610
Description
Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Flow CytometrySpecifications
Cyclin E1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
CCNE1/2460 | |
Flow Cytometry | |
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) (Uniprot: IDP24864) | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
898 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction