Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin E1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500129FR
Description
Cyclin E1 Polyclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Western BlotSpecifications
Cyclin E1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
Partial residue sequence of Cyclin E1. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers | |
898 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction