Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin E1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727261 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7261 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7261 Supplier Proteintech Group Inc Supplier No. 115541AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are expressed only in tumors but not in the normal tissues, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.

Specifications

Antigen Cyclin E1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CCNE1
Gene Accession No. P24864, Q61457
Gene Alias CCNE, CCNE1, cyclin E1, G1/S specific cyclin E1
Gene Symbols CCNE1
Host Species Rabbit
Immunogen Cyclin E1 Fusion Protein Ag2110
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12447, 898
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.