Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin E2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299464AF532
Description
Cyclin E2 Polyclonal specifically detects Cyclin E2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Cyclin E2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Cyclin E2. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers | |
9134 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction