Learn More
Invitrogen™ Cyclin E2 Polyclonal Antibody

Description
PA1-21582 detects Cyclin E2 in Human samples.
Specifications
Specifications
| Antigen | Cyclin E2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CCNE2 |
| Gene Accession No. | O96020 |
| Gene Alias | Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 |
| Gene Symbols | CCNE2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.