Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclin E2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586642
Description
Purity is > 95% by SDS-PAGE.
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
Cyclin E2 | |
Polyclonal | |
Unconjugated | |
CCNE2 | |
Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12448, 9134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O96020, Q9Z238 | |
CCNE2 | |
Synthetic peptide, corresponding to amino acids 351-404 of Human Cyclin E2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction